Learn More
Brady™ Laminated Polyester Warning Safety Signs
Marca: Brady™ 250220
Dettagli aggiuntivi : Weight : 0.01200kg
Specifica
ISO 7010, RoHS compliant using EU Directive 2002/95/EC, ROHS Environmental Compliance | |
1 Ea. | |
Laminated Polyester | |
200 mm | |
200 mm | |
Triangular | |
Warning Safety Sign |
Polyester | |
Permanent Acrylic | |
Pipe markers and safety signs | |
0.024 mm | |
-40°C to 110°C | |
Warning of industrial trucks | |
Black on Yellow |