Learn More
Laminin alpha 1 Antibody (775016), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4187CL1
Product Code. 30143517
  • € 987.00 / 0.10mL
Description
Laminin alpha 1 Monoclonal antibody specifically detects Laminin alpha 1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha 1
Monoclonal
CoraFluor 1
LAMA, LAMA1 laminin, alpha 1, S-LAM-alpha
E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391
0.1 mL
Primary
Human
Purified
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
775016
PBS
Mouse
Protein A or G purified
RUO
284217
Store at 4°C in the dark. Do not freeze.