missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 1 Antibody (775016), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4187K
This item is not returnable.
View return policy
Description
Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| Laminin alpha 1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284217 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry | |
| 775016 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| LAMA, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction