missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Laminin alpha-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119248
This item is not returnable.
View return policy
Description
Laminin alpha-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), ELISA
Laminin antibodies are widely used to label blood vessels and basement membranes.
Specifications
| Laminin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| LAMA1 | |
| LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 316758 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 0 | |
| LAMA1 | |
| Recombinant protein Laminin Alpha 1. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction