missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 4 Antibody (OTI5G8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71113AF350
This item is not returnable.
View return policy
Description
Laminin alpha 4 Monoclonal specifically detects Laminin alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Laminin alpha 4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LAMA4 | |
| Recombinant protein fragment corresponding to amino acids 25-270 of human Laminin alpha 4 (NP_001098676) produced in E.coli. | |
| 0.1 mL | |
| 3910.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5G8 | |
| Western Blot, Immunohistochemistry | |
| DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction