missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 202.00 - € 573.00
Specifications
| Antigen | Laminin beta 1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231094
|
Novus Biologicals
NBP3-38620-20ul |
20 μL |
€ 202.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227452
|
Novus Biologicals
NBP3-38620-100ul |
100 μL |
€ 573.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Laminin beta 1 Polyclonal antibody specifically detects Laminin beta 1 in Human,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Laminin beta 1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
| PBS (pH 7.3), 50% glycerol | |
| 3912 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CLM, cutis laxa with marfanoid phenotype, Laminin B1 chain, laminin subunit beta-1, laminin, beta 1, Laminin-1 subunit beta, Laminin-10 subunit beta, Laminin-12 subunit beta, Laminin-2 subunit beta, Laminin-6 subunit beta, Laminin-8 subunit beta, MGC142015 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1467-1786 of human Laminin beta 1 (NP_002282.2).,, Sequence:, EAKLRADEAKQSAEDILLKTNATKEKMDKSNEELRNLIKQIRNFLTQDSADLDSIEAVANEVLKMEMPSTPQQLQNLTEDIRERVESLSQVEVILQHSAADIARAEMLLEEAKRASKSATDVKVTADMVKEALEEAEKAQVAAEKAIKQADEDIQGTQNLLTSIESETAASEETLFNASQRISELERNVEELKRKAAQNSGEAEYIEKVVYTVKQSAEDVKKTLDGELDEKYKKVENLIAKKTEESADARRKAEMLQNEAKTLLAQANSKLQLLKDLERKYEDNQRYLEDKAQELARLEGEVRSLLKDISQKVAVYSTCL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title