missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Laminin beta-1 Monoclonal Antibody (CL2970)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA535004

Product Code. 17125673

  • € 638.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GPRCDKCTRG YSGVFPDCTP CHQCFALWDV IIAELTNRTH RFLEKAKALK ISGVIGPYRE TVDSVERKVS EIKDILAQSP AAEPLKNIGN LFEEAEKLIK DVTEMMAQVE VKLSDTTSQS NSTAKELDSL QTEAESLDNT VKELAEQLEF Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Laminin beta-1
Monoclonal
1.0 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P07942
LAMB1
Recombinant protein fragment of Human LAMB1 (Product #RP-102390)
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
CL2970
Unconjugated
LAMB1
C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015
Mouse
Protein A
RUO
3912
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.