Learn More
Invitrogen™ Laminin beta-2/gamma-1 Monoclonal Antibody (M046)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA547722
Product Code. 17836728
  • € 550.00 / 100µL
Description
Clone M046 detects native laminin isoforms that include both laminin beta2 and gamma1 subunits. In native immunoblots, the antibody detects laminin 521, laminin 221, and laminin 121, but does not detect lamin 332 and laminin 411. The antibody weakly detects laminin 511. This antibody can be used for immunocytochemical detection of laminin beta2/gamma1 subunits, and can also be used for detection of native laminin isoforms in immunoblots and ELISA.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin beta-2/gamma-1
Monoclonal
~1.0 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P11047, P55268
LAMB2, LAMC1
Clone (M046) was generated from a recombinant human laminin 521 protein that includes a heterotrimer of three subunits laminin alpha5, laminin beta2, and laminin gamma1 subunits.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot, Immunocytochemistry
M046
Unconjugated
LAMC1
AA408497; AW211941; B2e; basement membrane protein; Lama; Lamb2; Lamb-2; Lamc1; Laminin B1s chain; laminin B2 chain; laminin beta 2; laminin beta 2 chain; laminin chain B3; laminin chain beta 2; laminin gamma 1 chain; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin subunit gamma 1; laminin subunit gamma-1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1 subunit gamma; Laminin-10 subunit gamma; laminin-11 subunit beta; Laminin-11 subunit gamma; laminin-14 subunit beta; Laminin-15 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit beta; laminin-3 subunit gamma; Laminin-4 subunit beta; laminin-4 subunit gamma; Laminin-6 subunit gamma; laminin-7 subunit beta; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit beta; laminin-9 subunit gamma; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma
Mouse
Protein G
RUO
3913, 3915
-20°C, Avoid Freeze/Thaw Cycles