missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Laminin beta-2 Monoclonal Antibody (CL2976)
GREENER_CHOICE

Product Code. 30826278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30826278 Supplier Invitrogen™ Supplier No. MA562991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.

Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2976
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMB2
Gene Accession No. P55268
Gene Alias AW211941; Lamb2; Lamb-2; Laminin B1s chain; laminin chain B3; laminin chain beta 2; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin, beta 2; laminin, beta 2 (laminin S); laminin-11 subunit beta; Laminin-14 subunit beta; laminin-15 subunit beta; Laminin-3 subunit beta; laminin-4 subunit beta; Laminin-7 subunit beta; laminin-9 subunit beta; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-laminin subunit beta
Gene Symbols LAMB2
Host Species Mouse
Immunogen Recombinant protein corresponding to Human LAMB2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3913
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.