missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 3 Antibody (OTI8H5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71116IR
This item is not returnable.
View return policy
Description
Laminin beta 3 Monoclonal specifically detects Laminin beta 3 in Human samples. It is validated for Western Blot.
Specifications
| Laminin beta 3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LAMB3 | |
| Recombinant protein fragment corresponding to amino acids 940-1172 of human Laminin beta 3 (NP_000219) produced in E.coli. | |
| 0.1 mL | |
| 3914.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI8H5 | |
| Western Blot | |
| BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction