missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin gamma 1 Antibody (A5), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-47842PCP
This item is not returnable.
View return policy
Description
Laminin gamma 1 Monoclonal antibody specifically detects Laminin gamma 1 in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence, CyTOF
Specifications
| Laminin gamma 1 | |
| Monoclonal | |
| PerCP | |
| Q5VYE7, Q6NVY8 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Bovine | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| A5 | |
| PBS | |
| LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
| Murine EHS laminin preparation | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Tumor Suppressors | |
| 3915 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction