missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin gamma 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87719-25ul
This item is not returnable.
View return policy
Description
Laminin gamma 1 Polyclonal antibody specifically detects Laminin gamma 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Laminin gamma 1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ECREGFVGNRCDQCEENYFYNRSWPGCQECPACYRLVKDKVADHRVKLQELESLIANLGTGDEMVTDQAFEDRLKEAEREVMDLLREAQDVKDVDQNLMDRLQRVNNTLSSQISRLQNIRNTIEETGNLAEQARAHVENTERLIEIASR | |
| 25 μL | |
| Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Tumor Suppressors | |
| 3915 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction