missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin S/Laminin beta 2 Antibody (CL2976) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42386PCP
This item is not returnable.
View return policy
Description
Laminin S/Laminin beta 2 Monoclonal antibody specifically detects Laminin S/Laminin beta 2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Laminin S/Laminin beta 2 | |
| Monoclonal | |
| PerCP | |
| Laminin B1s chain, laminin S, laminin subunit beta-2, laminin, beta 2 (laminin S), Laminin-11 subunit beta, Laminin-14 subunit beta, Laminin-15 subunit beta, Laminin-3 subunit beta, Laminin-4 subunit beta, Laminin-7 subunit beta, LAMSLaminin-9 subunit beta, S-LAM beta, S-laminin subunit beta | |
| This antibody was developed using a recombinant protein derived from P55268, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL2976 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3913 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction