Learn More
LAMP-1/CD107a Antibody (508921), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB4800S
Product Code. 30062713
  • € 642.00 / 0.10mL
Description
LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAMP-1/CD107a
Monoclonal
Alexa Fluor 750
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
508921
Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Mouse myeloma cell line NS0-derived recombinant human LAMP1/CD107a, Ala28-Asn380, Accession # P11279
0.1 mL
Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites
3916
Store at 4C in the dark.
IgG2b