missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgr6 Antibody (918719) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8458M
This item is not returnable.
View return policy
Description
Lgr6 Monoclonal antibody specifically detects Lgr6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Lgr6 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 918719 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631 | |
| NS0 mouse myeloma cell line transfected with human Lgr6, Met1-Val967, Accession # Q9HBX8 | |
| 0.1 mL | |
| GPCR | |
| 59352 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction