missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-1/CD107a Antibody (H4A3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25183G
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| LAMP-1/CD107a | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LAMP1 | |
| Human PBMC | |
| 0.1 mL | |
| Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
| 3916 | |
| Human, Mouse, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| H4A3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The mouse monoclonal antibody H4A3 recognizes CD107a, an approximately 100-120 kDa glycoprotein expressed mainly on lysosomal, but also on the plasma membrane. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction