missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-2/CD107b Antibody (743320), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6228B
This item is not returnable.
View return policy
Description
LAMP-2/CD107b Monoclonal antibody specifically detects LAMP-2/CD107b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| LAMP-2/CD107b | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 743320 | |
| Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP2, LAMP-2, LAMP2A, LAMP-2A, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein 2 | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP2/CD107b, Leu29-Phe375, Accession # P13473 | |
| 0.1 mL | |
| Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| 3920 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction