missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP-2/CD107b Antibody (H4B4), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22217PECY55
This item is not returnable.
View return policy
Description
LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human, Mouse, Chicken, Rat (Negative) samples. It is validated for Flow Cytometry, Knockout Validated.
Specifications
| LAMP-2/CD107b | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein | |
| Mouse | |
| 45 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Chicken, Rat (Negative) | |
| IgG1 κ |
| Flow Cytometry, Immunoassay | |
| H4B4 | |
| Flow Cytometry, Knockout Validated | |
| P13473, P13473 | |
| LAMP2 | |
| LAMP-2/CD107b Antibody (H4B4) was made using a human adherent spleen cells. | |
| Protein G purified | |
| Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
| 3920.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction