missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP1, Mouse anti-Human, Clone: 6E2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16404.50uL
This item is not returnable.
View return policy
Specifications
| LAMP1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10 mM sodium azide). | |
| CD107a/LAMPA/LGP120 | |
| Mouse | |
| RUO | |
| 3916 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 6E2 | |
| Immunocytochemistry (1:2000) Immunofluorescence (1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:10000) The optimal working dilution should be determined by the end user. | |
| lysosomal-associated membrane protein 1 | |
| LAMP1 | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction