missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMP2 Monoclonal Antibody (M3/84)

Rat Monoclonal Antibody
€ 306.00 - € 606.00
Specifications
| Antigen | LAMP2 |
|---|---|
| Clone | M3/84 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Cytotoxicity Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15304141
![]() |
Invitrogen™
MA517861 |
100 μg |
€ 306.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15314141
|
Invitrogen™
MA517862 |
300 μg |
€ 606.00
300µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAMP2 Monoclonal Antibody for Western Blot, Flow, IP, CT
LAMP2 (Lysosome-associated membrane glycoprotein 2, CD107b) is a member of a family of membrane glycoproteins. LAMP2 provides selectins with carbohydrate ligands, and may play a role in tumor cell metastasis, and have a function in the protection, maintenance, and adhesion of the lysosome. Prior to posttranslational modification, LAMP2 is approximately 46 kDa polypeptide. Mature, functional LAMP2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of about 100-130 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. LAMP2 is used to monitor a very selective form of autophagy called chaperone-mediated autophagy. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. The LAMP protein has 3 variant forms: LAMP-2A, -2B, and 2C. LAMP2, together with CD107a/LAMP-1, is a major constituent of lysosomal membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes, and LAMP2 is an important regulator in successful phagosomal maturation. LAMP2 deficiency causes an accumulation of autophagosomes in many tissues leading to cardiomyopathy and myopathy (Danon's disease).Specifications
| LAMP2 | |
| 1 mg/mL | |
| Cytotoxicity Assay, Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.02% sodium azide | |
| P17047 | |
| 16784 | |
| Immunoadsorbent purified mouse macrophage glycoprotein fraction. | |
| Antibody |
| M3/84 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Mouse | |
| LAMP2 | |
| CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 | |
| LAMP2 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title