missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMTOR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572955
This item is not returnable.
View return policy
Description
LAMTOR3 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF
This gene encodes a scaffold protein that functions in the extracellular signal-regulated kinase (ERK) cascade. The protein is localized to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. Studies of the orthologous gene in mouse indicate that it regulates late endosomal traffic and cell proliferation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 13.
Specifications
| LAMTOR3 | |
| Polyclonal | |
| Unconjugated | |
| LAMTOR3 | |
| AW556229; hypothetical protein LOC553597; lamtor3; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 3; late endosomal/lysosomal adaptor, MAPK and MTOR activator 3; Map2k1ip1; MAPBP; MAPK scaffold protein 1; MAPKSP1; MEK binding partner 1; MEK partner 1; MEK-binding partner 1; mitogen activated protein, binding protein; mitogen-activated protein kinase kinase 1 interacting protein 1; mitogen-activated protein kinase kinase 1-interacting protein 1; Mitogen-activated protein kinase scaffold protein 1; mitogen-activated protein kinase scaffold protein 1; ragulator complex protein LAMTOR3; Mp1; PRO0633; PRO2783; Ragulator complex protein LAMTOR3; Ragulator3; zgc:110207 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56692, 8649 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.08 mg/mL | |
| PBS with 0.05% sodium azide | |
| O88653, Q9UHA4 | |
| LAMTOR3 | |
| A genomic peptide made to an internal region of the human MAP2K1IP1/MAPKSP1 protein (within residues 20-180). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction