missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAMTOR4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578680

Product Code. 16327374

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: HeLa, SKNSH Predicted Reactivity: Mouse (95%), Rat (96%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

LAMTOR4 is a part of the Ragulator complex it is involved in amino acid sensing and activation of mTORC1, a signaling complex promoting cell growth in response to growth factors, energy levels, and amino acids. LAMTOR4 is activated by amino acids through a mechanism involving the lysosomal V-ATPase, the Ragulator functions as a guanine nucleotide exchange factor activating the small GTPases Rag. LAMTOR4 is activated Ragulator and Rag GTPases function as a scaffold recruiting mTORC1 to lysosomes where it is in turn activated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAMTOR4
Polyclonal
Unconjugated
LAMTOR4
0910001L09Rik; AV006840; C7orf59; Lamtor4; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 4; late endosomal/lysosomal adaptor, MAPK and MTOR activator 4; ragulator complex protein LAMTOR4; Ragulator complex protein LAMTOR4, N-terminally processed; RGD1309735; UPF0539 protein C7orf59; UPF0539 protein C7orf59 homolog
Rabbit
Antigen affinity chromatography
RUO
389541
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
2.74 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Q0VGL1
LAMTOR4
Full length human LAMTOR4 recombinant protein.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.