missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LANA2 Antibody (CM-A807), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-167H
This item is not returnable.
View return policy
Description
LANA2 Monoclonal specifically detects LANA2 in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| LANA2 | |
| Monoclonal | |
| HRP | |
| LANA 2, LANA-2, Latency-associated nuclear antigen 2, vIRF 3, vIRF3, vIRF-3 | |
| Recombinant LANA2 fusion protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Chromatin Research, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| This is specific for KSHV LANA2 (residues 143-310 epitope recognition). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| CM-A807 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction