missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanatoside C, MedChemExpress
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Lanatoside C is a cardiac glycoside, can be used in the treatment of congestive heart failure and cardiac arrhythmia.Lanatoside C has an IC50 of 0.19 μM for dengue virus infection in HuH-7 cells. Lanatoside C can effectively inhibit all four serotypes of dengue virus, flavivirus Kunjin, alphavirus Chikungunya, Sindbis virus and the human enterovirus 71.
Specifications
Specifications
| Chemical Name or Material | Lanatoside C |
| CAS | 17575-22-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C49H76O20 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (50.76 mM; Need ultrasonic) ∣Ethanol : 2 mg/mL (2.03 mM; Need ultrasonic) |
| SMILES | O[C@]([C@@](CC[C@@]1([H])[C@@]2(CC[C@H](O[C@@](O[C@H](C)[C@H]3O[C@@](O[C@H](C)[C@H]4O[C@@](O[C@H](C)[C@H]5O[C@]([C@@H]([C@@H](O)[C@@H]6O)O)([H])O[C@@H]6CO)([H])C[C@@H]5OC(C)=O)([H])C[C@@H]4O)([H])C[C@@H]3O)C1)C)([H])[C@]2([H])C[C@H]7O)(CC[C@@H]8C(CO9)=CC9=O)[C@]78C |
| Molecular Weight (g/mol) | 985.12 |
| Formula Weight | 985.12 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction