missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Langerin/CD207 Antibody (LGRN/7430), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20734R
This item is not returnable.
View return policy
Description
Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Langerin/CD207 | |
| Monoclonal | |
| DyLight 550 | |
| CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin | |
| Recombinant fragment (around aa50-250) of human Langerin/CD207 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| metabolism, Signal Transduction | |
| 50489 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| LGRN/7430 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction