missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LANPL Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77227MFV450
This item is not returnable.
View return policy
Description
LANPL Polyclonal antibody specifically detects LANPL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LANPL | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| acidic (leucine-rich) nuclear phosphoprotein 32 family, member E, LANP-like protein, LANP-LLANPLacidic leucine-rich nuclear phosphoprotein 32 family member E, leucine-rich acidic nuclear protein like, MGC5350 | |
| Antibody was raised with a synthetic peptide corresponding to amino acids close to carboxy terminus of human PHAP III. The immunogen is located within amino acids 200 - 250 of PHAP III. Amino Acid Squence: GLSYLMKEEIQDEE | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 81611 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction