Learn More
Lanthanum Standard: La @ 1000 μg/mL in 5% HNO3, VHG™
Product Code. 17977769
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977769 500 mL 500mL
17929299 50 mL 50mL
17958379 100 mL 100mL
17968879 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17977769 Supplier VHG Labs Supplier No. VHGPLAN500
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Standard: La @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Lanthanum
CAS 7439-91-0
Methods ICP/ICP-MS
Analyte or Component Names Lanthanum
Shipping Condition Room Temperature
Molecular Formula La
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym La
InChI Formula InChI=1S/La
SMILES [La]
IUPAC Name lanthanum
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 138.906
Quantity 500 mL
Formula Weight 138.906 g/mol