missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Eu-anti-GST Antibody
LanthaScreen™ Eu-anti-GST Antibody
Brand: Invitrogen PV5595
This item is not returnable.
View return policy
Description
LanthaScreen™ europium-labeled anti-GST goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
Specifications
| GST | |
| 0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
| 150kDa | |
| RUO | |
| Primary | |
| Europium-labeled anti-GST antibody is supplied in HEPES-buffered saline. Store at -20°C. | |
| Liquid | |
| Microplate Reader | |
| GST | |
| LanthaScreen |
| Kinase Assay | |
| Europium | |
| 1 mg | |
| Pharma and Biopharma | |
| 1 Kit(s) | |
| 150 kDa | |
| 340⁄665 | |
| Dry Ice | |
| TR-FRET |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction