Learn More
Invitrogen™ LanthaScreen™ Tb-anti-ubiquitin-FK2 antibody
The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin.
Brand: Invitrogen™ PV4734
Description
The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen™ cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled anti-ubiquitin-FK2 antibody in HEPES buffered saline. Store at -20°C. |
|
| Human | |
| End Point | |
| Ubiquitin |
| 1 mg | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| LanthaScreen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.