missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LanthaScreen™ Tb-Streptavidin, 1 mg/mL Kit

Product Code. 10574803
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
This item is not returnable. View return policy

Product Code. 10574803

Brand: Invitrogen™ PV3966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LanthaScreen™ Tb-Streptavidin, 1 mg/mL Kit

As part of the LanthaScreen™ TR-FRET Toolbox of assay reagents, LanthaScreen™ Tb-labeled streptavidin is a generic reagent that can be used to easily label biotinylated biomolecules with terbium for use in TR-FRET assays. Although the terbium chelate donor can be directly attached to a biomolecule such as a protein or peptide, the Tb-labeled streptavidin can be indirectly attached to a target molecule through a biotin-mediated association with Tb-labeled streptavidin.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate Tb (Terbium)
Quantity 1 mg
Content And Storage LanthaScreen™ Tb-Streptavidin is supplied at 1 mg/ml and contains the streptavidin in HEPES buffered saline, 400 μl of the fluorescein-labeled, biotinylated peptide for use as a positive control, and 100 ml of TR-FRET Dilution Buffer. Store components as indicated in the product Certificate of Analysis (-20°C, or +20-30°C).
Shipping Condition Dry Ice
Form Liquid
Product Line LanthaScreen
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Tb (Terbium)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.