missing translation for 'onlineSavingsMsg'
Learn More

LAP (Latency Associated Peptide) Monoclonal Antibody (7F6)

LAP (Latency Associated Peptide) Monoclonal Antibody (7F6)

Brand:  Invitrogen MA517186

Product Code. 15358150

  • € 497.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LAP (Latency Associated Peptide) Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAP (Latency Associated Peptide)
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P01137
TGFB1
Purified recombinant fragment of human TGFb1 (amino acids: 62-195) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
7F6
Unconjugated
TGFB1
CED; DPD1; LAP; latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGFB; Tgfb1; Tgfb-1; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1; transforming growth factor-beta 1
Mouse
Protein G
RUO
7040
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.