missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP (TGF-beta 1) Antibody (7F6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22114B
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TGFB1 | |
| Purified recombinant fragment of human TGF beta 1 (amino acids 62-197) expressed in E. coli. [UniProt# P01137] | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific | |
| 7040 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 7F6 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction