missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP (TGF-beta 1) Antibody (860206), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7666H
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| LAP (TGF-beta 1) | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 860206 | |
| Flow Cytometry, CyTOF-ready | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse LAP (TGF-beta 1), Met1-Ser390, Accession # P04202 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction