Learn More
LAP1 Antibody (RL13), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2737B
This item is not returnable.
View return policy
Description
LAP1 Monoclonal antibody specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LAP1 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| RL13 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| lamina-associated polypeptide 1B, torsin A interacting protein 1 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| 0.2 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 208263 | |
| Store at 4°C in the dark. | |
| IgG1 |