missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16360965
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16360965 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16360965 Supplier Invitrogen™ Supplier No. PA584013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EATSVQQKVN FSEEGETEED DQDSSHSSVT TVKARSRDSD ESGDKTTRSS SQYIESFWQS SQSQNFTAHD KQRSVLSSGY QKTPQEWAPQ TARIRTRMQ Highest antigen sequence indentity to the following orthologs: Rat - 47%, Mouse - 46%.

Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOR1AIP1
Gene Accession No. Q5JTV8
Gene Alias lamina-associated polypeptide 1B; Lamina-associated polypeptide 1C; Lamin-associated protein 1B; LAP1; LAP1B; Lap1c; Tor1aip1; torsin 1A interacting protein 1; torsin A interacting protein 1; torsin-1A-interacting protein 1
Gene Symbols TOR1AIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOR1AIP1. Recombinant protein control fragment (Product #RP-99776).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.