Learn More
LAPTM4A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30627-1-AP-150UL
 View more versions of this product
Product Code. 18005305
  • € 410.00 / 150µL
Description
The LAPTM4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LAPTM4A. This antibody recognizes human, mouse antigen. The LAPTM4A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
LAPTM4A
1000 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
HUMORF13, KIAA0108, LAPTM4, LAPTM4A, MBNT, Mtrp
150 μL
9741
Store at -20°C. Stable for one year after shipment.