Learn More
LAPTM4A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30627-1-AP-150UL
Description
The LAPTM4A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LAPTM4A. This antibody recognizes human, mouse antigen. The LAPTM4A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| LAPTM4A | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| HUMORF13, KIAA0108, LAPTM4, LAPTM4A, MBNT, Mtrp | |
| 150 μL | |
| 9741 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |