missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAPTM4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18895-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize all the 3 isoforms of LAPTM4B.
LAPTM4B is a multi-pass membrane protein. It belongs to the LAPTM4/LAPTM5 transporter family. LAPTM4b has active role in disease progression of maligt cells and is involved in cell proliferation and multidrug resistance. The genetic polymorphism of LAPTM4B is a potential risk factor for the development of colon cancer and gastric cancer. The research results also indicated that LAPTM4B may be a clinically useful prognostic indicator for ovarian carcinoma and may play a role in human hepatocellular carcinoma.Specifications
| LAPTM4B | |
| Polyclonal | |
| Unconjugated | |
| LAPTM4B | |
| LAPTM4B, LAPTM4beta, LC27 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114128, 315047, 55353 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U1W4, Q86VI4, Q91XQ6 | |
| Laptm4b | |
| LAPTM4B Fusion Protein Ag13521 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction