missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAR/PTPRF Antibody (W7C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-90995
This item is not returnable.
View return policy
Description
LAR/PTPRF Monoclonal specifically detects LAR/PTPRF in Human, Primate samples. It is validated for Flow Cytometry.
Specifications
| LAR/PTPRF | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry 1 to 4 μg/mL | |
| LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F | |
| WERI-RB-1 retinoblastoma cells | |
| 0.1 mg | |
| Primary | |
| Human, Primate | |
| Purified |
| Flow Cytometry | |
| W7C6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5792 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction