Learn More
Invitrogen™ LAR4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143989
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human Jurkat whole cell, human K562 whole cell. IHC: human appendiceal adenocarcinoma tissue, human liver cancer tissue. ICC/IF: A431 cell. Flow: THP-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The LAR4B gene, more accurately referred to as LARP4B (La Ribonucleoprotein Domain Family Member 4B), is associated with RNA-binding proteins that are crucial for various aspects of RNA metabolism, including RNA stabilization, translation, and cellular stress responses. LARP4B belongs to the La-related protein family, which plays significant roles in maintaining RNA stability and enhancing translation efficiency of certain mRNAs. It has been identified as a target in cancer research due to its involvement in tumor biology, particularly regulating tumor suppressive pathways and interacting with microRNA networks. Studies have shown that LARP4B expression is downregulated in prostate cancer tissues, indicating its potential role as a tumor suppressor. Moreover, it is linked with liver cancer and survival rates, reflecting its importance in cellular growth and proliferation. The regulation of LARP4B and its interactions with other cellular components underline its potential therapeutic implications for cancer treatment.
Specifications
| LAR4B | |
| Polyclonal | |
| Unconjugated | |
| LARP4B | |
| A630096F19; AI256361; D13Wsu64e; KIAA0217; La ribonucleoprotein 4B; La ribonucleoprotein domain family member 4B; La ribonucleoprotein domain family member 5; La ribonucleoprotein domain family, member 4B; La ribonucleoprotein domain family, member 5; la-related protein 4B; La-related protein 5; LARP4B; Larp5; RGD1310713 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23185 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q92615 | |
| LARP4B | |
| E.coli-derived human LARP4B recombinant protein (Position: R241-K684). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.