Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250μL and larger.
Brand:  Hamilton™ 7732-10
Product Code. 12336309
  • € 404.00 / Pack of 6
Explore more special offers
Description
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers