Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250μL and larger.
Brand: Hamilton™ 7732-10
This item is not returnable.
View return policy
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style and lengths
- Autoclavable
- For large hubs