Learn More
Gilac™ Large Volume Semicircular Container
Click to view available options
Capacity (Metric):
170 L
220 L
Unit Size:
Each
Description
- Large volume semicircular container.
- Ideal for culinary preparations.
- Extremely durable.
- Resistance to chemical products.
- Tough and very strong.
- Compatible with big dough container trolleys.
- Standing work height.
- Volume in litres is equivalent to the container filled to the brim.
Specifications
Specifications
| Type | Storage Tote |
| Material | HDPE |
| Color | White |
| Shape | Semi-circular |
| Capacity (English) | 58.1 gal. |
| Capacity (Metric) | 220 L |