missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LARP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518653
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
LARP2 encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
Specifications
| LARP2 | |
| Polyclonal | |
| Unconjugated | |
| LARP1B | |
| 1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 55132 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q659C4 | |
| LARP1B | |
| Synthetic peptide sequence (QGVLHIPKKDLTDE) corresponding to the internal amino acids of LARP2 (aa 553-567). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction