missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22254DL594
This item is not returnable.
View return policy
Description
LARP4 Polyclonal antibody specifically detects LARP4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| LARP4 | |
| Polyclonal | |
| Western Blot | |
| c-Mpl binding protein, DKFZp686E039, La ribonucleoprotein domain family member 4, La ribonucleoprotein domain family, member 4, la-related protein 4, MGC74631, PP13296 | |
| The immunogen this antibody was made to, maps to a region between residue 674 and 724 of human La Ribonucleoprotein Domain Family, Member 4 using the numbering given in entry NP_443111.4 (GeneID 113251). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 113251 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction