missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LARP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555437
This item is not returnable.
View return policy
Description
Immunogen sequence: DSGVPQNTGM KNEKTANREE CRTQEKVNAT GPQFVSGVIV KIISTEPLPG RKQVRDTLAA ISEVLYVDLL EGDTECHA Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 56%.
LARP7 gene ontology annotations related to this gene include RNA processing.
Specifications
| LARP7 | |
| Polyclonal | |
| Unconjugated | |
| LARP7 | |
| ALAZS; C330027G06Rik; D3Wsu161e; DKFZP564K112; HDCMA18P; La ribonucleoprotein 7, transcriptional regulator; La ribonucleoprotein domain family member 7; La ribonucleoprotein domain family, member 7; la-related protein 7; la-related protein 7; LOW QUALITY PROTEIN: la-related protein 7; Larp7; PIP7S; P-TEFb-interaction protein for 7SK stability; Unknown (protein for IMAGE:8171460); Unknown (protein for IMAGE:8281412) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4G0J3 | |
| LARP7 | |
| Recombinant protein corresponding to Human LARP7. Recombinant protein control fragment (Product #RP-91861). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction