missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | LARS |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227489
|
Novus Biologicals
NBP3-35479-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232830
|
Novus Biologicals
NBP3-35479-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LARS Polyclonal antibody specifically detects LARS in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| LARS | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| cytoplasmic leucyl-tRNA synthetase, FLJ21788, hr025Cl, HSPC192, KIAA1352, LARS1, leucine translase, leucine tRNA ligase 1, cytoplasmic, leucine-tRNA ligase, leucine--tRNA ligase, leucyl-tRNA synthetase, leucyl-tRNA synthetase, cytoplasmic, LEURS, LEUS, LRS, PIG44 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human LARS (NP_064502.9).,, Sequence:, MAERKGTAKVDFLKKIEKEIQQKWDTERVFEVNASNLEKQTSKGKYFVTFPYPYMNGRLHLGHTFSLSKCEFAVGYQRLKGKCCLFPFGLHCTGMPIKACADKLKREIELYGCPPDFPDEEEEEEETSVKTEDIIIKDKAKGKKSKAAAKAGSSKYQWGIMKSLGLSDEEIVKFSEAEHWLDYFPPLAIQDLKRMGLKVDWRRSFITTDVNPYYDSFVRWQFLTLRERNKIKFGKRYTIYSPKDGQPCMDHDRQTGEGVGPQEYTLLKLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 51520 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title