missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 515.00
Specifications
| Antigen | LARS2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LARS2 Polyclonal specifically detects LARS2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| LARS2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 23395 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GCSWRSGAKVEQKYLRQWFIKTTAYAKAMQDALADLPEWYGIKGMQAHWIGDCVGCHLDFTLKVHGQATGEKLTAYTATPE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| EC 6.1.1, EC 6.1.1.4, KIAA0028leucine tRNA ligase 2, mitocondrial, leucine translase, Leucine--tRNA ligase, leucyl-tRNA synthetase 2, mitochondrial, LEURS, MGC26121, probable leucyl-tRNA synthetase, mitochondrial | |
| LARS2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title