missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAS2 Antibody (OTI2B2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72452F
This item is not returnable.
View return policy
Description
LAS2 Monoclonal antibody specifically detects LAS2 in Human samples. It is validated for Western Blot
Specifications
| LAS2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 162681 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI2B2 | |
| Western Blot | |
| C18orf54, chromosome 18 open reading frame 54, hypothetical protein LOC162681, LAS2, MGC33382 | |
| Full length human recombinant protein of human C18orf54(NP_775800) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction