Learn More
Lasalocid A Sodium Salt (Solution in acetonitrile 0.1mg/ml), TRC
Specifications
Specifications
| Chemical Name or Material | Lasalocid A sodium salt |
| CAS | 25999-20-6 |
| Molecular Formula | C34 H53 O8 . Na |
| Quantity | 5 mL |
| Synonym | 2,3-Cresotic acid, 6-[7-[5-ethyl-5-(5-ethyltetrahydro-5-hydroxy-6-methyl-2H-pyran-2-yl)tetrahydro-3-methyl-2-furyl]-4-hydroxy-3,5-dimethyl-6-oxononyl]-, monosodium salt, (-)- (8CI), Benzoic acid, 6-[(3R,4S,5S,7R)-7-[(2S,3S,5S)-5-ethyl-5-[(2R,5R,6S)-5-ethyltetrahydro-5-hydroxy-6-methyl-2H-pyran-2-yl]tetrahydro-3-methyl-2-furanyl]-4-hydroxy-3,5-dimethyl-6-oxononyl]-2-hydroxy-3-methyl-, monosodium salt (9CI), Benzoic acid, 6-[7-[5-ethyl-5-(5-ethyltetrahydro-5-hydroxy-6-methyl-2H-pyran-2-yl)tetrahydro-3-methyl-2-furanyl]-4-hydroxy-3,5-dimethyl-6-oxononyl]-2-hydroxy-3-methyl-, monosodium salt, [2R-[2α[2S*(3R*,4S*,5S*,7R*),3S*,5S*],5α,6β]]-, Avatec, Bovatec, Lasalocid sodium, Lasalocid sodium salt, MT 2007, NSC 243046, Ro 2-2985, Sodium lasalocid, Sodium lasalocid A |
| InChI Formula | InChI=1S/C34H54O8.Na/c1-9-25(31-21(6)18-34(11-3,42-31)26-16-17-33(40,10-2)23(8)41-26)30(37)22(7)28(35)19(4)12-14-24-15-13-20(5)29(36)27(24)32(38)39;/h13,15,19,21-23,25-26,28,31,35-36,40H,9-12,14,16-18H2,1-8H3,(H,38,39);/q;+1/p-1/t19-,21+,22+,23+,25+,26-,28+,31+,33-,34+;/m1./s1 |
| SMILES | [Na+].CC[C@H]([C@H]1O[C@@](CC)(C[C@@H]1C)[C@H]2CC[C@](O)(CC)[C@H](C)O2)C(=O)[C@@H](C)[C@@H](O)[C@H](C)CCc3ccc(C)c(O)c3C(=O)[O-] |
| IUPAC Name | sodium;6-[(3R,4S,5S,7R)-7-[(2S,3S,5S)-5-ethyl-5-[(2R,5R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-3-methyloxolan-2-yl]-4-hydroxy-3,5-dimethyl-6-oxononyl]-2-hydroxy-3-methylbenzoate |
| Molecular Weight (g/mol) | 612.77 |
| Formula Weight | 612.3638 |
| Show More |