Learn More
Lasalocid sodium, MedChemExpress
Product Code. 16478658
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478658 10 mg 10mg
16474537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16478658 Supplier Medchem Express Supplier No. HYB1071A
MedChemExpress Lasalocid sodium (Lasalocid-A sodium) treatment led to an increase in cell wall thickness, whilst the quantity and sugar composition of the cell wall remained unchanged in BY-2 cells.
Lasalocid sodium (Lasalocid-A sodium) treatment led to an increase in cell wall thickness, whilst the quantity and sugar composition of the cell wall remained unchanged in BY-2 cells.
Specifications
Chemical Name or Material Lasalocid sodium
Physical Form Solid
CAS 25999-20-6
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C34H53NaO8
For Use With (Application) COVID-19-immunoregulation
Synonym Lasalocid-A sodiumIonophore X-537A sodiumAntibiotic X-537A sodium
Solubility Information DMSO : 100 mg/mL (163.19 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O[Na])C1=C(CC[C@@H](C)[C@H](O)[C@H](C)C([C@@H]([C@H]2O[C@@]([C@H]3CC[C@](O)(CC)[C@H](C)O3)(CC)C[C@@H]2C)CC)=O)C=CC(C)=C1O
Molecular Weight (g/mol) 612.77
Formula Weight 612.77
Percent Purity 97.0%
Grade Research
Show More Show Less
Research purposes only