missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody (661002), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB63341APCCY7
This item is not returnable.
View return policy
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Flow Cytometry
Specifications
| LAT | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 661002 | |
| Flow Cytometry | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| E. coli-derived recombinant human LAT, His30-Gln113, Accession # O43561 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 27040 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction